Fmoc-Gly-Gly-Gly-Gly 97%

Supplier: Ambeed
Warning

A155819-100MG A155819-1G A155819-250MG A155819-5G A155819-25G
76973-304EA 7 USD
76973-304 76973-306 76973-308 77549-980 77881-221
Fmoc-Gly-Gly-Gly-Gly 97%
Fmoc-Gly-Gly-Gly-Gly

Precaución: for Research Use Only

Formula: C₂₃H₂₄N₄O₇
MDL Number: MFCD18427731
CAS Number: 1001202-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR