4-Hydroxy-2-aminobenzoxazol 95%

Supplier: Ambeed
Warning

A249393-5G A249393-250MG
76976-656EA 460 USD
76976-656 77672-222
4-Hydroxy-2-aminobenzoxazol 95%
4-Hydroxy-2-aminobenzoxazol

Precaución: for Research Use Only

Formula: C₇H₆N₂O₂
MDL Number: MFCD18819810
CAS Number: 98549-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR