6-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine 95%

Supplier: Ambeed
Warning

A455600-1G A455600-5G
76979-508EA 175 USD
76979-508 77572-438
6-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine 95%
6-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₃ClIN₃
MW: 279.466 g/mol
MDL Number: MFCD18250995
CAS Number: 1260672-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR