(E)-2-(3-Chlorostyryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A606939-1G A606939-250MG A606939-5G
76980-534EA 154 USD
76980-534 76980-536 76980-538
(E)-2-(3-Chlorostyryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
(E)-2-(3-Chlorostyryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₁₈BClO₂
MW: 264.56 g/mol
MDL Number: MFCD08276867
CAS Number: 871125-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR