4-Chloro-β-styrylboronic acid pinacol ester 98+%

Supplier: Ambeed
Warning

Synonyms: (E)-2-(4-Chlorostyryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A914363-1G A914363-250MG A914363-5G A914363-25G
76981-938EA 52 USD
76981-938 76981-940 76981-942 77882-818
4-Chloro-β-styrylboronic acid pinacol ester 98+%
4-Chloro-beta-styrylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BClO₂
MW: 264.56 g/mol
MDL Number: MFCD15144834
CAS Number: 223919-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR