PP242 ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)-1H-indol-5-ol, Torkinib, 2-[4-Amino-1-(1-methylethyl)-1H-pyrazolo[3,4-d]pyrimidin-3-yl]-1H-indol-5-ol

A228542-10MG A228542-1G A228542-1MG A228542-5MG A228542-250MG A228542-50MG
76976-034EA 37.14 USD
76976-034 76976-036 76976-038 76976-040 77672-022 77672-024
PP242 ≥98%
PP242

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₆O
MW: 308.3 g/mol
MDL Number: MFCD12196869
CAS Number: 1092351-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR