2-Aminoquinoline-7-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A724119-100MG
A724119-1G
A724119-250MG
A724119-50MG
76981-246EA
135.8
USD
76981-246
76981-248
76981-250
76981-252
2-Aminoquinoline-7-carboxylic acid 95%
2-Aminoquinoline-7-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₈N₂O₂ MW: 188.18 g/mol |
MDL Number:
MFCD11557272 CAS Number: 1092287-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...