3-Methyl-1H-indazole-5-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-1H-indazol-5-yl-5-boronic acid

A537815-1G A537815-250MG A537815-5G
76980-142EA 170.81 USD
76980-142 76980-144 77569-044
3-Methyl-1H-indazole-5-boronic acid 97%
3-Methyl-1H-indazole-5-boronic acid

Precaución: For research use only.

Formula: C₈H₉BN₂O₂
MW: 175.9800 g/mol
MDL Number: MFCD12028643
CAS Number: 1245816-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR