1-(Quinolin-2-yl)ethanone 95%

Supplier: Ambeed
Warning

A727953-250MG A727953-1G
76981-264EA 56 USD
76981-264 77573-338
1-(Quinolin-2-yl)ethanone 95%
1-(Quinolin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₉NO
MDL Number: MFCD08062664
CAS Number: 1011-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR