Methyl-2-bromoimidazo[1,2-a]pyridine-6-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-bromoimidazo[1,2-a]pyridine-6-carboxylate

A189591-100MG A189591-1G A189591-250MG
76974-810EA 14 USD
76974-810 76974-812 76974-814
Methyl-2-bromoimidazo[1,2-a]pyridine-6-carboxylate 98%
Methyl 2-bromoimidazo[1,2-a]pyridine-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
MW: 255.0700 g/mol
MDL Number: MFCD13195435
CAS Number: 1042141-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR