3-Acetyl-5-bromo-2-fluoropyridine 98%

Supplier: Ambeed
Warning

A110777-1G A110777-250MG A110777-25G A110777-5G
76970-758EA 15 USD
76970-758 76970-760 76970-762 76970-764
3-Acetyl-5-bromo-2-fluoropyridine 98%
3-Acetyl-5-bromo-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₇H₅BrFNO
MDL Number: MFCD12024449
CAS Number: 1111637-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR