Ethyl-2-cyano-3,3-diphenylacrylate 98%

Supplier: Ambeed
Warning

A185076-1000G A185076-100G A185076-25G A185076-500G
76974-566EA 98 USD
76974-566 76974-568 76974-570 76974-572 76974-574
Ethyl-2-cyano-3,3-diphenylacrylate 98%
Ethyl-2-cyano-3,3-diphenylacrylate

Precaución: for Research Use Only

Formula: C₁₈H₁₅NO₂
MDL Number: MFCD00027364
CAS Number: 5232-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR