2,6-Dichloro-3-methyl-5-nitropyridine 95%

Supplier: Ambeed
Warning

A137040-10G A137040-1G A137040-250MG
76972-382EA 600.47 USD
76972-382 76972-384 76972-386
2,6-Dichloro-3-methyl-5-nitropyridine 95%
2,6-Dichloro-3-methyl-5-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O₂
MW: 207.0100 g/mol
MDL Number: MFCD17014968
CAS Number: 58596-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR