ZM 447439 ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
ZM-447439, N-(4-((6-Methoxy-7-(3-morpholinopropoxy)quinazolin-4-yl)amino)phenyl)benzamide
A107524-10MG
A107524-1MG
A107524-5MG
76970-502EA
42.44
USD
76970-502
76970-504
76970-506
ZM 447439 ≥98%
ZM 447439
Precaución: for Research Use Only
Formula:
C₂₉H₃₁N₅O₄ MW: 513.6 g/mol |
MDL Number:
MFCD08703125 CAS Number: 331771-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...