Etravirine 98%

Supplier: Ambeed
Warning

Synonyms: 4-((6-Amino-5-bromo-2-((4-cyanophenyl)amino)pyrimidin-4-yl)oxy)-3,5-dimethylbenzonitrile

A403297-10MG A403297-1G A403297-1MG A403297-50MG A403297-5MG A403297-100MG A403297-250MG A403297-5G
76979-084EA 12.74 USD
76979-084 76979-086 76979-088 76979-090 76979-092 77613-236 77613-238 77672-972
Etravirine 98%
Etravirine

Precaución: for Research Use Only

Formula: C₂₀H₁₅BrN₆O
MW: 435.29 g/mol
MDL Number: MFCD09837879
CAS Number: 269055-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR