2-Bromo-6-(trifluoromethyl)phenol 95%

Supplier: Ambeed
Warning

A362806-1G A362806-250MG A362806-5G
76978-612EA 181.42 USD
76978-612 76978-614 77551-050
2-Bromo-6-(trifluoromethyl)phenol 95%
2-Bromo-6-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O
MW: 241.005 g/mol
MDL Number: MFCD13194490
CAS Number: 2844-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR