5-Bromo-2-formylbenzonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-cyanobenzaldehyde

A138930-1G A138930-250MG A138930-5G
76972-502EA 129 USD
76972-502 76972-504 76972-506
5-Bromo-2-formylbenzonitrile 95%
5-Bromo-2-formylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrNO
MW: 210.027 g/mol
MDL Number: MFCD09261030
CAS Number: 523977-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR