9-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-4,7-dioxanonanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 9-(Fmoc-amino)-4,7-dioxanonanoic acid, Fmoc-NH-PEG2-CH2CH2COOH, Fmoc-9-amino-4,7-dioxanonanoic acid, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10-trioxa-4-azatridecan-13-oic acid, 3-[2-[2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]ethoxy]ethoxy]propionic acid, 3-[2-[2-(Fmoc-amino)ethoxy]ethoxy]propionic acid

A221766-10G A221766-1G A221766-250MG A221766-5G A221766-25G A221766-100G
76975-854EA 62 USD
76975-854 76975-856 76975-858 77550-436 77568-136 77612-152
9-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-4,7-dioxanonanoic acid 98%
9-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-4,7-dioxanonanoic acid

Precaución: For research use only.

Formula: C₂₂H₂₅NO₆
MDL Number: MFCD08064306
CAS Number: 872679-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR