2-Cyclopropyl-6-hydroxypyrimidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A271643-1G A271643-250MG
76977-192EA 20.16 USD
76977-192 76977-194
2-Cyclopropyl-6-hydroxypyrimidine-4-carboxylic acid 97%
2-Cyclopropyl-6-hydroxypyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₈N₂O₃
MW: 180.16 g/mol
MDL Number: MFCD19689702
CAS Number: 858956-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR