2-Bromo-4-nitropyridine-N-oxide 98%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-4-nitropyridine N-oxide, 2-Bromo-4-nitropyridine 1-oxide

A614510-1G A614510-25G A614510-5G
76980-568EA 10.61 USD
76980-568 76980-570 76980-572
2-Bromo-4-nitropyridine-N-oxide 98%
2-Bromo-4-nitropyridine-N-oxide

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₃
MW: 218.99 g/mol
MDL Number: MFCD00160743
CAS Number: 52092-43-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR