3-Formyl-1H-indazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A108609-1G A108609-250MG
76970-568EA 116 USD
76970-568 77880-880
3-Formyl-1H-indazole-5-carboxylic acid 97%
3-Formyl-1H-indazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₃
MDL Number: MFCD06656900
CAS Number: 885519-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR