2-(4-Bromophenyl)-1,1-diphenylethylene 95%

Supplier: Ambeed
Warning

A420211-1G A420211-250MG A420211-5G
76979-234EA 43.5 USD
76979-234 76979-236 76979-238
2-(4-Bromophenyl)-1,1-diphenylethylene 95%
2-(4-Bromophenyl)-1,1-diphenylethylene

Precaución: for Research Use Only

Formula: C₂₀H₁₅Br
MW: 335.24 g/mol
MDL Number: MFCD01790865
CAS Number: 18648-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR