(S)-4-(4-(5-(Aminomethyl)-2-oxooxazolidin-3-yl)phenyl)morpholin-3-one 97%

Supplier: Ambeed
Warning

A294991-100MG A294991-1G A294991-250MG A294991-5G A294991-25G
76977-628EA 9 USD
76977-628 76977-630 76977-632 76977-634 77881-807
(S)-4-(4-(5-(Aminomethyl)-2-oxooxazolidin-3-yl)phenyl)morpholin-3-one 97%
(S)-4-(4-(5-(Aminomethyl)-2-oxooxazolidin-3-yl)phenyl)morpholin-3-one

Precaución: for Research Use Only

Formula: C₁₄H₁₇N₃O₄
MW: 291.30 g/mol
MDL Number: MFCD11977666
CAS Number: 446292-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR