4-Methoxy-3-(3-methoxypropoxy)benzaldehyde 98+%

Supplier: Ambeed
Warning

A467445-1G A467445-5G
76979-612EA 33.95 USD
76979-612 76979-614
4-Methoxy-3-(3-methoxypropoxy)benzaldehyde 98+%
4-Methoxy-3-(3-methoxypropoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄
MW: 224.25 g/mol
MDL Number: MFCD11113215
CAS Number: 172900-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR