4'-Methoxy-3'-methylacetophenone 97%
Supplier: Ambeed
|
Warning
|
A110996-100MG
A110996-1G
A110996-250MG
A110996-5G
76970-774EA
5
USD
76970-774
76970-776
76970-778
76970-780
4'-Methoxy-3'-methylacetophenone 97%
4'-Methoxy-3'-methylacetophenone
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₂ |
MDL Number:
MFCD01673307 CAS Number: 10024-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...