N-(5-Bromo-2-methylphenyl)-3-cyanobenzamide 95%

Supplier: Ambeed
Warning

A489875-100MG A489875-250MG
76979-784EA 21.22 USD
76979-784 76979-786
N-(5-Bromo-2-methylphenyl)-3-cyanobenzamide 95%
N-(5-Bromo-2-methylphenyl)-3-cyanobenzamide

Precaución: for Research Use Only

Formula: C₁₅H₁₁BrN₂O
MDL Number: MFCD12719083
CAS Number: 1157457-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR