4-Methoxy-2-nitro-1-(trifluoromethyl)benzene 95%

Supplier: Ambeed
Warning

A847010-1G A847010-5G A847010-25G
76981-788EA 14 USD
76981-788 76981-790 77882-768
4-Methoxy-2-nitro-1-(trifluoromethyl)benzene 95%
4-Methoxy-2-nitro-1-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₃
MDL Number: MFCD11848539
CAS Number: 25889-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR