3-Bromo-4,5-dichloropyridine 97%

Supplier: Ambeed
Warning

A151063-100MG A151063-250MG A151063-1G A151063-5G A151063-25G
76973-116EA 7 USD
76973-116 76973-118 77592-836 77592-838 77881-171
3-Bromo-4,5-dichloropyridine 97%
3-Bromo-4,5-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrCl₂N
MDL Number: MFCD17014545
CAS Number: 1001056-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR