1H-Pyrrolo[2,3-b]pyridin-6-ol 95%

Supplier: Ambeed
Warning

A124754-100MG A124754-1G A124754-250MG
76971-782EA 54.11 USD
76971-782 76971-784 76971-786
1H-Pyrrolo[2,3-b]pyridin-6-ol 95%
1H-Pyrrolo[2,3-b]pyridin-6-ol

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.1353 g/mol
MDL Number: MFCD08272226
CAS Number: 55052-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR