Dabigatran Etexilate 98%

Supplier: Ambeed
Warning

Synonyms: Pradaxa, Hexyl 2-(4-(((5-((3-ethoxy-3-oxopropyl)(pyridin-2-yl)carbamoyl)-1-methyl-1H-benzo[d]imidazol-2-yl)methyl)amino)benzylidene)hydrazinecarboxylate, Ethyl 3-(2-(((4-(N-((hexyloxy)carbonyl)carbamimidoyl)phenyl)amino)methyl)-1-methyl-N-(pyridin-2-yl)-1H-benzo[d]imidazole-5-carboxamido)propanoate, BIBR 953, (E)-ethyl 3-(2-(((4-(N'-((hexyloxy)carbonyl)carbamimidoyl)phenyl)amino)methyl)-1-methyl-N-(pyridin-2-yl)-1H-benzo[d]imidazole-5-carboxamido)propanoate

A249359-100MG A249359-1G A249359-250MG A249359-50MG A249359-25G A249359-5G
76976-648EA 6.37 USD
76976-648 76976-650 76976-652 76976-654 77568-416 77612-408
Dabigatran Etexilate 98%
Dabigatran Etexilate

Precaución: For research use only.

Formula: C₃₄H₄₁N₇O₅
MW: 627.73 g/mol
MDL Number: MFCD16038312
CAS Number: 211915-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR