Cbz-D-Homoserine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Cbz-D-Hse-OH
A107413-1G
A107413-250MG
76970-486EA
49.87
USD
76970-486
76970-488
Cbz-D-Homoserine 98%
Cbz-D-Homoserine
Precaución: for Research Use Only
Formula:
C₁₂H₁₅NO₅ MW: 253.25 g/mol |
MDL Number:
MFCD12545887 CAS Number: 41088-85-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...