3-(Difluoromethyl)aniline hydrochloride 95%

Supplier: Ambeed
Warning

A679360-1G A679360-250MG A679360-5G
76980-998EA 175 USD
76980-998 76981-000 76981-002
3-(Difluoromethyl)aniline hydrochloride 95%
3-(Difluoromethyl)aniline hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈ClF₂N
MDL Number: MFCD16547606
CAS Number: 161363-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR