1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d][1,2,3]triazole 98%

Supplier: Ambeed
Warning

A790689-100MG A790689-1G A790689-250MG A790689-5G
76981-604EA 248.26 USD
76981-604 76981-606 76981-608 76981-610
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d][1,2,3]triazole 98%
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₁₃H₁₈BN₃O₂
MW: 259.11 g/mol
MDL Number: MFCD19689515
CAS Number: 1396753-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR