1-tert-Butyl-1H-pyrazole-4-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(tert-Butyl)-1H-pyrazole-4-carboxylic acid
A349247-100MG
A349247-1G
A349247-250MG
A349247-5G
76978-498EA
46.68
USD
76978-498
76978-500
76978-502
76978-504
1-tert-Butyl-1H-pyrazole-4-carboxylic acid 98%
1-tert-Butyl-1H-pyrazole-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₁₂N₂O₂ MW: 168.19 g/mol |
MDL Number:
MFCD09934934 CAS Number: 950858-65-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...