AT9283 98%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclopropyl-3-(3-(5-(morpholinomethyl)-1H-benzo[d]imidazol-2-yl)-1H-pyrazol-4-yl)urea

A335492-10MG A335492-1MG A335492-50MG A335492-5MG
76978-326EA 273.72 USD
76978-326 76978-328 76978-330 76978-332
AT9283 98%
AT9283

Precaución: for Research Use Only

Formula: C₁₉H₂₃N₇O₂
MW: 381.4 g/mol
MDL Number: MFCD12031513
CAS Number: 896466-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR