GW2580 98%

Supplier: Ambeed
Warning

A161762-10MG A161762-1MG
76973-620EA 29 USD
76973-620 76973-622
GW2580 98%
GW2580

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₄O₃
MDL Number: MFCD12024701
CAS Number: 870483-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR