5,8-Dichloropyrido[2,3-d]pyridazine 98%

Supplier: Ambeed
Warning

A399035-1G A399035-250MG A399035-5G A399035-25G
76979-034EA 37 USD
76979-034 76979-036 76979-038 77882-064
5,8-Dichloropyrido[2,3-d]pyridazine 98%
5,8-Dichloropyrido[2,3-d]pyridazine

Precaución: for Research Use Only

Formula: C₇H₃Cl₂N₃
MDL Number: MFCD00193165
CAS Number: 703-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR