Ethyl 2-(7-methoxynaphthalen-1-yl)acetate 97%
Supplier: Ambeed
|
Warning
|
A188372-1G
A188372-25G
A188372-5G
76974-750EA
9.55
USD
76974-750
76974-752
76974-754
Ethyl 2-(7-methoxynaphthalen-1-yl)acetate 97%
Ethyl 2-(7-methoxynaphthalen-1-yl)acetate
Precaución: for Research Use Only
Formula:
C₁₅H₁₆O₃ MW: 244.28 g/mol |
MDL Number:
MFCD12756805 CAS Number: 6836-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...