2-(Methylsulfonyl)ethanamine 95%

Supplier: Ambeed
Danger

A235096-1G A235096-250MG A235096-25G A235096-5G
76976-308EA 20 USD
76976-308 76976-310 76976-312 76976-314
2-(Methylsulfonyl)ethanamine 95%
2-(Methylsulfonyl)ethanamine

Precaución: for Research Use Only

Formula: C₃H₉NO₂S
MDL Number: MFCD01862553
CAS Number: 49773-20-8
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR