5-Bromo-1,3-difluoro-2-methylbenzene 98%

Supplier: Ambeed
Warning

A147544-100G A147544-10G A147544-1G A147544-25G A147544-5G
76972-908EA 578.2 USD
76972-908 76972-910 76972-912 76972-914 76972-916
5-Bromo-1,3-difluoro-2-methylbenzene 98%
5-Bromo-1,3-difluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
MDL Number: MFCD07368697
CAS Number: 179617-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR