5,7-Difluoro-3,4-dihydronaphthalen-1(2H)-one 98%

Supplier: Ambeed
Warning

A365097-100MG A365097-250MG
76978-644EA 22 USD
76978-644 76978-646
5,7-Difluoro-3,4-dihydronaphthalen-1(2H)-one 98%

Precaución: for Research Use Only

Formula: C₁₀H₈F₂O
MDL Number: MFCD08544417
CAS Number: 110931-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR