(5-Fluoropyrimidin-2-yl)acetic acid 98%

Supplier: Ambeed
Warning

A595218-100MG A595218-250MG
76980-456EA 133 USD
76980-456 76980-458
(5-Fluoropyrimidin-2-yl)acetic acid 98%
(5-Fluoropyrimidin-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₂
MDL Number: MFCD13189601
CAS Number: 1196145-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR