(2-Aminothiazol-4-yl)methanol 95%

Supplier: Ambeed
Warning

A143698-1G A143698-250MG A143698-5G
76972-716EA 77 USD
76972-716 76972-718 76972-720
(2-Aminothiazol-4-yl)methanol 95%
(2-Aminothiazol-4-yl)methanol

Precaución: for Research Use Only

Formula: C₄H₆N₂OS
MDL Number: MFCD08275702
CAS Number: 51307-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR