4-Bromo-3-methyl-1H-pyrazol-5(4H)-one 95%

Supplier: Ambeed
Warning

A137456-1G A137456-5G
76972-400EA 44 USD
76972-400 76972-402
4-Bromo-3-methyl-1H-pyrazol-5(4H)-one 95%
4-Bromo-3-methyl-1H-pyrazol-5(4H)-one

Precaución: for Research Use Only

Formula: C₄H₅BrN₂O
MDL Number: MFCD16620159
CAS Number: 51395-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR