Boc-L-Arg(Z)2-OH 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Arg(Cbz)₂-OH, Boc-Arg(Z)2-OH

A111047-1G A111047-250MG A111047-5G
76970-792EA 24 USD
76970-792 76970-794 77549-150
Boc-L-Arg(Z)2-OH 97%
Boc-L-Arg(Z)2-OH

Precaución: for Research Use Only

Formula: C₂₇H₃₄N₄O₈
MDL Number: MFCD00236824
CAS Number: 51219-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR