2-(N-Boc-piperidin-3-yl)-1H-benzoimidazole 96%

Supplier: Ambeed
Warning

A763691-1G A763691-250MG
76981-444EA 34 USD
76981-444 76981-446
2-(N-Boc-piperidin-3-yl)-1H-benzoimidazole 96%
2-(N-Boc-piperidin-3-yl)-1H-benzoimidazole

Precaución: for Research Use Only

Formula: C₁₇H₂₃N₃O₂
MDL Number: MFCD17214278
CAS Number: 1229000-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR