Methyl-6-iodonicotinate 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-iodonicotinate

A556973-1G A556973-250MG
76980-256EA 118.83 USD
76980-256 76980-258
Methyl-6-iodonicotinate 96%
Methyl-6-iodonicotinate

Precaución: for Research Use Only

Formula: C₇H₆INO₂
MW: 263.03 g/mol
MDL Number: MFCD12546492
CAS Number: 173157-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR