4-Bromo-2-methyl-1H-benzimidazole 97%

Supplier: Ambeed
Warning

A340734-100MG A340734-1G A340734-250MG
76978-396EA 14 USD
76978-396 76978-398 77672-740
4-Bromo-2-methyl-1H-benzimidazole 97%
4-Bromo-2-methyl-1H-benzimidazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MDL Number: MFCD11521322
CAS Number: 20223-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR