Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate 95%

Supplier: Ambeed
Warning

A261709-10G A261709-25G A261709-5G A261709-100G
76976-974EA 19 USD
76976-974 76976-976 76976-978 76976-980 77881-709
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate 95%
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
MDL Number: MFCD00087563
CAS Number: 51145-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR