L-Lysinamide dihydrochloride 97%

Supplier: Ambeed
Warning

A124861-250MG A124861-1G A124861-5G
76971-800EA 49 USD
76971-800 77881-003 77881-004
L-Lysinamide dihydrochloride 97%
L-Lysinamide dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₇Cl₂N₃O
MDL Number: MFCD00058287
CAS Number: 51127-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR